2-Aminoindan

Leverantör: Thermo Scientific
Varning

SB01761DA SB01761EA
MAYBSB01761DAEA 0 SEK
MAYBSB01761DA MAYBSB01761EA
2-Aminoindan
2-Aminoindan
Formel: C₉H₁₁N
MDL Number: MFCD00082597
CAS nummer: 2975-41-9
EINECS: 221-021-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR