5,6-Dimethoxy-1-indanone ≥97%
Leverantör: Thermo Scientific
Synonyms: 5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one
B25559.06 B25559.14
B25559.06EA 1725 SEK
B25559.06 B25559.14
5,6-Dimethoxy-1-indanone ≥97%
5,6-Dimethoxy-1-indanone
Purity: 97% CAS No.: 2107-69-9 Molecular Formula: C11H12O3 Formula Weight: 192.21 Melting Point: 118-120° MDL No.: MFCD00003790
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
Kokpunkt: 302 °C (1013 hPa)
Smältpunkt: 118…120 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003790
CAS nummer: 2107-69-9
EINECS: 218-287-8