5-Methoxy-1-indanone 98%
Leverantör: Thermo Scientific
Varning
179670050 179670100
ACRO179670050EA 1500 SEK
ACRO179670050 ACRO179670100
5-Methoxy-1-indanone 98%
5-Methoxy-1-indanone
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokpunkt: 145…150 °C (0,2 mmHg)
Smältpunkt: 107…111 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003789
CAS nummer: 5111-70-6
EINECS: 225-838-6
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Green-brown Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 111 °C
GC ≥97.5 %