Selendisulfid ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Selenium sulphide, Selenium(IV) sulphide
68104.06 68104.18 68104.30
68104.06EA 865 SEK
68104.06 68104.18 68104.30
Selendisulfid ≥97%
Selendisulfid
Purity: 97% CAS No.: 7488-56-4 Molecular Formula: SeS2 Formula Weight: 143.09 Physical Form: Powder Melting Point: 100° Boiling Point: dec. Merck Reference: 14,8436 UN No.: 2657 MDL No.: MFCD00011227
Formel: S₂Se
MW: 143,09 g/mol
Kokpunkt: Dec.
Smältpunkt: <100 °C
Densitet: 0,03 g/cm³ (–65 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011227
CAS nummer: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1,II
Merck Index: 14,8436