Fosforpentoxid ISO, ACS, Reag. Ph. Eur. zA, Supelco®
Leverantör: Merck
|
Fara
|
Synonyms:
Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
MQ200
1.00570.0100
1.00570.0500
1.00570.0100EA
961
SEK
1.00570.0100
1.00570.0500
Fosforpentoxid ISO, ACS, Reag. Ph. Eur. zA, Supelco®
Fosforpentoxid
Formel:
P₂O₅ MW: 141.94 g/mol Smältpunkt: 562 Densitet: 2.253 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011440 CAS nummer: 1314-56-3 EINECS: 215-236-1 UN: 1807 ADR: 8 II Merck Index: 13,07441 |