Fosforpentoxid ≥98% ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
10524.18 10524.36 10524.57
10524.18EA 660 SEK
10524.18 10524.36 10524.57
Fosforpentoxid ≥98% ACS
Fosforpentoxid
Purity: 98% min CAS No.: 1314-56-3 Molecular Formula: P2O5 Formula Weight: 141.94 Physical Form: Powder Density: 2.39 Appearance: White Odour: Weak Melting Point: 340° Boiling Point: 360° subl. Solubility: Decomposes in water. Soluble in H2SO4. Insoluble in acetone and NH3 Merck Reference: 14,7355 UN No.: 1807 MDL No.: MFCD00011440
As a drying and dehydrating agent, as a condensing agent in organic synthesis, in sugar refining, as a laboratory reagent, and in fire extinguishing
Formel: P₂O₅
MW: 141,94 g/mol
Kokpunkt: 360 °C (subl.)
Smältpunkt: 340 °C
Densitet: 2,39
Storage Temperature: Omgivande
MDL Number: MFCD00011440
CAS nummer: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 14,7355