Kiselgel 99.9%, pulver 0,5 µm
Leverantör: Thermo Scientific
Varning
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
L16985.14 L16985.22
L16985.14EA 1700 SEK
L16985.14 L16985.22
Kiselgel 99.9%, pulver 0,5 µm
Kiselgel
Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C
Smältpunkt: 1710 °C
Densitet: 2,0
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493