Kiselgel ≥99.99%
Leverantör: Thermo Scientific
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
36346.14 36346.22
36346.14EA 2375 SEK
36346.14 36346.22
Kiselgel ≥99.99%
Kiselgel
Purity: 99.99% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 3-12mm Fused Lump Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C
Smältpunkt: 1710 °C
Densitet: 2,2…2.6
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493