Kiselgel ≥99.999%, Puratronic®
Leverantör: Thermo Scientific
Fara
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
10856.14 10856.22
10856.14EA 5750 SEK
10856.14 10856.22
Kiselgel ≥99.999%, Puratronic®
Kiselgel
Purity: 99.999% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C
Smältpunkt: 1710 °C
Densitet: 2,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 7631-86-9
EINECS: 238-878-4
Merck Index: 14,8493