Kiselgel, Pore size: 60 Å Ca, ~0.1% 230-400 mesh, high purity, Supelco®
Leverantör: Merck
M-Clarity™ quality level = MQ200
60752-1KG
60752-5KG
60752-1KGEA
2610
SEK
60752-1KG
60752-5KG
Kiselgel, Pore size: 60 Å Ca, ~0.1% 230-400 mesh, high purity, Supelco®
Kiselgel
Formel:
SiO₂ Storage Temperature: Omgivande |
MDL Number:
MFCD00011232 CAS nummer: 112926-00-8 |