Kiselgel, Pore size: 60 Å 35-60 mesh, high purity, Supelco®
Leverantör: Merck
M-Clarity™ quality level = MQ100
60742-1KG 60742-25KG
60742-1KGEA 1550 SEK
60742-1KG 60742-25KG
Kiselgel, Pore size: 60 Å 35-60 mesh, high purity, Supelco®
Kiselgel
Formel: SiO₂
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8ab4cc0b12527585b6641fb374f4805b5c48959&vid=3f8d94354f149f3abd134327fe45819930dc3924&prid=web&clid=SAPDAM