orto-Borsyra, EMPROVE® ESSENTIAL BP, Ph. Eur., NF suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
1.00160.5000 1.00160.9050 1.00160.9025
1.00160.5000EA 6475 SEK
1.00160.5000 1.00160.9050 1.00160.9025
orto-Borsyra, EMPROVE® ESSENTIAL BP, Ph. Eur., NF suitable for use as excipient, SAFC®
orto-Borsyra
Formel: B(OH)₃
Kokpunkt: 185 °C (1013 hPa)
Smältpunkt: 160 °C
Densitet: 1,44 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011337
CAS nummer: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM