orto-Borsyra ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
A10896.0C A10896.0E A10896.36
A10896.0CEA 7125 SEK
A10896.0C A10896.0E A10896.36
orto-Borsyra ≥99%
orto-Borsyra
Purity: 99+% CAS No.: 10043-35-3 Molecular Formula: BH3O3 Formula Weight: 61.83 Flash Point: None Odour: Odourless pH: 3.8-4.8 Melting Point: ca 185° dec. Beilstein Reference: 1697939 Merck Reference: 14,1336 MDL No.: MFCD00011337
Formel: B(OH)₃
Kokpunkt: 185 °C (1013 hPa)
Smältpunkt: ∼ 185 °C (dec.)
Densitet: 1,435
Storage Temperature: Omgivande
MDL Number: MFCD00011337
CAS nummer: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 14,1336
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM