Dithalliumtrioxid 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Thallium (III) oxide
A17601.18 A17601.09
A17601.18EA 4350 SEK
A17601.18 A17601.09
Dithalliumtrioxid 96%
Dithalliumtrioxid
Purity: 96% CAS No.: 1314-32-5 Molecular Formula: Tl2O3 Formula Weight: 456.74 Odour: Odourless Melting Point: 717° Boiling Point: 875° Merck Reference: 14,9268 UN No.: 1707 MDL No.: MFCD00011276
Formel: O₃Tl₂
MW: 456.76 g/mol
Kokpunkt: 875 °C
Smältpunkt: 717
Densitet: 9.65
Storage Temperature: Omgivande
MDL Number: MFCD00011276
CAS nummer: 1314-32-5
EINECS: 215-229-3
UN: 1707
ADR: 6.1 II
Merck Index: 12,09404