Dibortrioxid ≥99.98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Boron oxide, Boric anhydride
89964.36 89964.22
89964.36EA 6775 SEK
89964.36 89964.22
Dibortrioxid ≥99.98%
Dibortrioxid
Purity: 99.98% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: Powder Density: 2.46 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315
Formel: B₂O₃
Kokpunkt: 1860 °C
Smältpunkt: ∼450 °C
Densitet: 2,46
Storage Temperature: Omgivande
MDL Number: MFCD00011315
CAS nummer: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337