Tallium(I)nitrat 99.5%
Leverantör: Thermo Scientific
Fara
Synonyms: Thallium nitrate
265590250 265591000
ACRO265590250EA 1575 SEK
ACRO265590250 ACRO265591000
Tallium(I)nitrat 99.5%
Tallium(I)nitrat
Formel: NO₃Tl
MW: 266,39 g/mol
Kokpunkt: 433 °C (760 mmHg)
Smältpunkt: 206 °C
Densitet: 1,273 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011280
CAS nummer: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Total trace metal impurities ≤5000 ppm