Cysteamine ≥95.0%

Leverantör: TCI
Fara

Synonyms: 2-Aminoethanethiol

A0648-500G A0648-25G
TCIAA0648-500GEA 3700 SEK
TCIAA0648-500G TCIAA0648-25G
Cysteamine ≥95.0%
Cysteamine
Formel: C₂H₇NS
Storage Temperature: Kylskåp
MDL Number: MFCD00008196
CAS nummer: 60-23-1
EINECS: 200-463-0
Merck Index: 14,02779
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a55bfc80f3fcb7b12f93fd0642f2cda306a316&vid=87a55bfc80f3fcb7b12f93fd0642f2cda306a316&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 94.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR