Indium(III) chloride 99.995%
Leverantör: Thermo Scientific
Fara
Synonyms: Indium Trichloride Chloride, Indium trichloride
193190100 193190500 193192500
ACRO193190100EA 1700 SEK
ACRO193190100 ACRO193190500 ACRO193192500
Indium(III) chloride 99.995%
Indium(III) chloride
Formel: Cl₃In
MW: 221,18 g/mol
Kokpunkt: 800 °C (1013 hPa)
Smältpunkt: 586 °C
Densitet: 4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011058
CAS nummer: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 13,04980
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Total trace metal impurities ≤0.005 %