2-Mercapto-2-thiazoline, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: 2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
M-Clarity™ quality level = MQ200
8.21111.0100 8.21111.0500 8.21111.9010
8.21111.0100EA 585 SEK
8.21111.0100 8.21111.0500 8.21111.9010
2-Mercapto-2-thiazoline, Sigma-Aldrich®
2-Mercapto-2-thiazoline
Formel: C₃H₅NS₂
Storage Temperature: Omgivande
MDL Number: MFCD00126013
CAS nummer: 96-53-7
EINECS: 202-512-1
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Solubility (20 °C) insoluble
Melting Point 101 - 106 °C
Molar Mass 119.21 g/mol
pH 6 - 7 (10 g/l, H2O, 20 °C)