5,5-Diphenylhydantoin, Sigma-Aldrich®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ200
8.20534.0005 8.20534.0250
8.20534.0005EA 1160 SEK
8.20534.0005 8.20534.0250
5,5-Diphenylhydantoin, Sigma-Aldrich®
5,5-Diphenylhydantoin
Formel: C₁₅H₁₂N₂O₂
MW: 252,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005264
CAS nummer: 57-41-0
EINECS: 200-328-6
Order Now
Specifikation testresultat
Solubility 0.032 g/l (22 °C) (experimental)
Melting Point 292 - 295 °C
Molar Mass 252.27 g/mol
Bulk Density 400 - 450 kg/m3
Ignition Point 550 °C