N-tert-Butylbenzylamine, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine
MQ200
8.18660.0005 8.18660.0250
8.18660.0005EA 494 SEK
8.18660.0005 8.18660.0250
N-tert-Butylbenzylamine, Sigma-Aldrich®
N-tert-Butylbenzylamine
Formel: C₁₁H₁₇N
MW: 163.26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008798
CAS nummer: 3378-72-1
EINECS: 222-179-6
UN: 2735
ADR: 8 II
Order Now
Specifikation testresultat
Solubility 2 g/l (20 °C)
Melting Point -30 - -25 °C
Molar Mass 163.26 g/mol
Boiling Point 218 - 220 °C
Vapor Pressure 0.07 hPa (53 °C)
Flash Point 90 °C
Refractive Index 1.4968 (20 °C, 589 nm)
Density 0.90 g/cm3 (20 °C)