Pyridine-2-carboxaldoxime, Sigma-Aldrich®
Leverantör: Merck
|
Varning
|
Synonyms:
2-Pyridinecarbaldehyde oxime, 2-Pyridinealdoxime
M-Clarity™ quality level = MQ200
8.07466.0025
8.07466.0100
8.07466.0025EA
480
SEK
8.07466.0025
8.07466.0100
Pyridine-2-carboxaldoxime, Sigma-Aldrich®
Pyridine-2-carboxaldoxime
Formel:
C₆H₆N₂O MW: 122,13 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00006354 CAS nummer: 873-69-8 EINECS: 212-849-6 |
Specifikation testresultat
Solubility | 15 g/l (20 °C) |
Melting Point | 110 - 112 °C |
Molar Mass | 122.12 g/mol |
Bulk Density | 400 - 500 kg/m3 |
pH | 7 (13 g/l, H2O, 20 °C) |