4H-3,1-Benzoxazin-2,4(1H)-dion, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: 1H-Benzo[d][1,3]oxazine-2,4-dione
M-Clarity™ quality level = MQ200
8.04784.0100 8.04784.0500
8.04784.0100EA 639 SEK
8.04784.0100 8.04784.0500
4H-3,1-Benzoxazin-2,4(1H)-dion, Sigma-Aldrich®
4H-3,1-Benzoxazin-2,4(1H)-dion
Formel: C₈H₅NO₃
Storage Temperature: Omgivande
MDL Number: MFCD00006700
CAS nummer: 118-48-9
EINECS: 204-255-0
Order Now
Specifikation testresultat
Solubility 0.3 g/l (20 °C)
Melting Point 230 - 232 °C
Molar Mass 163.13 g/mol
Bulk Density 600 kg/m3
Boiling Point 350 °C (1013 hPa)
Flash Point 308 °C
Ignition Point 600 °C