3,4-Dihydro-2H-pyran, Sigma-Aldrich®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ200
8.02971.0100 8.02971.0250
8.02971.0100EA 557 SEK
8.02971.0100 8.02971.0250
3,4-Dihydro-2H-pyran, Sigma-Aldrich®
3,4-Dihydro-2H-pyran
Formel: C₅H₈O
MW: 84,12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006558
CAS nummer: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II
Order Now
Specifikation testresultat
Solubility 20 g/l (20 °C)
Melting Point -70 °C
Molar Mass 84.12 g/mol
Boiling Point 89 °C
Vapor Pressure 120 hPa (20 °C)
Flash Point -9 °C
Refractive Index 1.441 (20 °C, 589 nm)
Explosion Limit 1.1 - 13.8 %(V)
Density 0.93 g/cm3 (20 °C)
pH 7 (5 g/l, H2O, 20 °C)
Ignition Point 240 °C