(±)-2,3-Epoxypropyl methacrylate Stabilised zS
Leverantör: Merck
Fara
Synonyms: Oxiran-2-ylmethyl methacrylate, Glycidyl methacrylate
Stabilization: Hydrochinonmonomethylether
8.00609.0250 8.00609.9025 8.00609.2500
8.00609.0250EA 665 SEK
8.00609.0250 8.00609.9025 8.00609.2500
(±)-2,3-Epoxypropyl methacrylate Stabilised zS
(±)-2,3-Epoxypropyl methacrylate
Formel: C₇H₁₀O₃
MW: 142.15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00005137
CAS nummer: 106-91-2
EINECS: 203-441-9
UN: 2922
ADR: 8 (6.1) III