Aluminiumklorid, vattenfri 99%, granular
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Aluminum chloride
A11892.0B A11892.30 A11892.0E
A11892.0BEA 1100 SEK
A11892.0B A11892.30 A11892.0E
Aluminiumklorid, vattenfri 99%, granular
Aluminiumklorid
Purity: 99% CAS No.: 7446-70-0 Molecular Formula: AlCl3 Formula Weight: 133.34 Odour: Pungent pH: 2,4 Melting Point: 190° subl. Merck Reference: 14,337 UN No.: 1726 MDL No.: MFCD00003422
Formel: AlCl₃
MW: 133.34 g/mol
Kokpunkt: 180
Smältpunkt: 190 °C (subl.)
Densitet: 2.44
Storage Temperature: Omgivande
MDL Number: MFCD00003422
CAS nummer: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 14,337