(R)-(-)-2-Chloromandelic acid ≥99%, ee 99%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
L19586.03 L19586.06 L19586.14
L19586.03EA 635 SEK
L19586.03 L19586.06 L19586.14
(R)-(-)-2-Chloromandelic acid ≥99%, ee 99%, ChiPros®
(R)-(-)-2-Chloromandelic acid
Purity: 99+% CAS No.: 52950-18-2 Molecular Formula: C8H7ClO3 Formula Weight: 186.60 Melting Point: 119-121° Beilstein Reference: 2693370
Formel: C₈H₇ClO₃
MW: 186,59 g/mol
Smältpunkt: 119…121 °C
Densitet: 1,48
Storage Temperature: Omgivande
MDL Number: MFCD00798429
CAS nummer: 52950-18-2
EINECS: 444-040-0