(S)-(-)-Hydroxypyrrolidine 99%, ee 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
L19498.03 L19498.06
L19498.03EA 3375 SEK
L19498.03 L19498.06
(S)-(-)-Hydroxypyrrolidine 99%, ee 99%
(S)-(-)-Hydroxypyrrolidine
Purity: 99% CAS No.: 100243-39-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 110°(230°F) Refractive Index: 1.4900 Melting Point: 15° Boiling Point: 108-110°/8mm Beilstein Reference: 4652606 Optical Rotation: -6.5° (c=3.5 in methanol) MDL No.: MFCD00192426
Formel: C₄H₉NO
MW: 87,12 g/mol
Kokpunkt: 108…110 °C (8 mmHg)
Smältpunkt: 15 °C
Densitet: 1,048
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00192426
CAS nummer: 100243-39-8