(±)-Allyl-2,3-epoxypropyl ether ≥97%
Leverantör: Thermo Fisher Scientific
Fara
L11634.22 L11634.36
L11634.22EA 815 SEK
L11634.22 L11634.36
(±)-Allyl-2,3-epoxypropyl ether ≥97%
(±)-Allyl-2,3-epoxypropyl ether
Purity: 97% CAS No.: 106-92-3 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 47°(134°F) Refractive Index: 1.4350 Odour: Ether-like Melting Point: -100° Boiling Point: 50-51°/15mm Beilstein Reference: 105871 UN No.: 2219 MDL No.: MFCD00005143
Formel: C₆H₁₀O₂
Kokpunkt: 50…51 °C (15 mmHg)
Smältpunkt: –100 °C
Densitet: 0,967
Flampunkt: 47 °C (134 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005143
CAS nummer: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III