(±)-2,3-Epoxypropyl methacrylate ≥97% stabilised
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Oxiran-2-ylmethyl methacrylate, Glycidyl methacrylate
Stabilization: Stabiliseras med hydrokinon Monometyltetraklordifenylmetan eter
L11133.18 L11133.30
L11133.18EA 565 SEK
L11133.18 L11133.30
(±)-2,3-Epoxypropyl methacrylate ≥97% stabilised
(±)-2,3-Epoxypropyl methacrylate
Stabilized with 100 ppm 4-methoxyphenol
Purity: 97% CAS No.: 106-91-2 Molecular Formula: C7H10O3 Formula Weight: 142.16 Flash Point: 83°(181°F) Refractive Index: 1.4500 Melting Point: -82° Boiling Point: 189° Beilstein Reference: 2506 UN No.: 2810 MDL No.: MFCD00005137
Formel: C₇H₁₀O₃
MW: 142.15 g/mol
Kokpunkt: 189 °C
Smältpunkt: –82 °C
Densitet: 1.078
Flampunkt: 83 °C (181 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005137
CAS nummer: 106-91-2
EINECS: 203-441-9
UN: 2810
ADR: 6.1 III