N,N'-Bis(tert-butyl)ethylenediamine 98%
Leverantör: Thermo Scientific
Fara
Synonyms: N1,N2-Di-tert-butylethane-1,2-diamine, N,N'-Di-tert-butylethylenediamine
L04972.06 L04972.14
L04972.06EA 1325 SEK
L04972.06 L04972.14
N,N'-Bis(tert-butyl)ethylenediamine 98%
N,N'-Bis(tert-butyl)ethylenediamine
Purity: 98% CAS No.: 4062-60-6 Molecular Formula: C10H24N2 Formula Weight: 172.32 Flash Point: 62°(143°F) Refractive Index: 1.4300 Boiling Point: 132°/100mm Beilstein Reference: 1738432 UN No.: 2735 MDL No.: MFCD00014996
Formel: C₁₀H₂₄N₂
MW: 172,31 g/mol
Kokpunkt: 132 °C (100 mmHg)
Densitet: 0,801
Flampunkt: 62 °C (143 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014996
CAS nummer: 4062-60-6
EINECS: 223-769-6
ADR: 8,II