N,N-Dimethylacetamide dimethyl acetal tech. 90% stabilised, Technical Grade
Leverantör: Thermo Scientific
Fara
Synonyms: (1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine
Stabilization: med metanol 5-10%
L01642.06 L01642.14
L01642.06EA 1250 SEK
L01642.06 L01642.14
N,N-Dimethylacetamide dimethyl acetal tech. 90% stabilised, Technical Grade
N,N-Dimethylacetamide dimethyl acetal
Purity: 90% CAS No.: 18871-66-4 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 8°(46°F) Refractive Index: 1.4120 Boiling Point: 118-120° Beilstein Reference: 1420865 UN No.: 1993 MDL No.: MFCD00008476
Formel: C₆H₁₅NO₂
MW: 133,19 g/mol
Kokpunkt: 118…120 °C
Densitet: 0,911
Flampunkt: 8 °C (46 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008476
CAS nummer: 18871-66-4
EINECS: 242-641-0
UN: 1993
ADR: 3,II