4-Hydroxypicolinic acid ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Hydroxypicolinic acid, 4-Hydroxy-2-pyridinecarboxylic acid, 4-Hydroxypyridine-2-carboxylic acid
H34262.03 H34262.MD
H34262.03EA 3650 SEK
H34262.03 H34262.MD
4-Hydroxypicolinic acid ≥97%
4-Hydroxypicolinic acid
Purity: 97% CAS No.: 22468-26-4 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Melting Point: ca 280° dec. MDL No.: MFCD00521155
Formel: C₆H₅NO₃
MW: 139,11 g/mol
Smältpunkt: ∼ 280 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00521155
CAS nummer: 22468-26-4