2-Hydroxypropyl methacrylate (mixture of isomers) ≥98% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester
Stabilization: Stabilised with 4-methoxyphenol (MEHQ) ∼ 0,02%
A17951.AK A17951.AK_BB A17951.0J A17951.AU
A17951.AKEA 830 SEK
A17951.AK ALFAA17951.AK_BB A17951.0J A17951.AU
2-Hydroxypropyl methacrylate (mixture of isomers) ≥98% stabilised
2-Hydroxypropyl methacrylate (mixture of isomers)
Stabilized with ca 0.02% 4-methoxyphenol
Purity: 98% CAS No.: 27813-02-1 Molecular Formula: CH2=C(CH3)CO2CH2CH(OH)CH3 Formula Weight: 144.17 Storage Temperature: 2°C to 8°C Flash Point: 97°(207°F) Refractive Index: 1.4480 Melting Point: 70°/1mm Boiling Point: 205-209° MDL No.: MFCD00004536
Formel: C₇H₁₂O₃
Kokpunkt: 205…209 °C
Smältpunkt: 70 °C/1mm
Densitet: 1,029
Flampunkt: 97 °C (207 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00004536
CAS nummer: 27813-02-1
EINECS: 248-666-3