Ferrocenecarboxylic acid ≥98%

Leverantör: Thermo Scientific
Varning

Synonyms: Ferrocene monocarboxylic acid

A16874.03 A16874.06
A16874.03EA 1475 SEK
A16874.03 A16874.06
Ferrocenecarboxylic acid ≥98%
Ferrocenecarboxylic acid

Purity: 98% CAS No.: 1271-42-7 Molecular Formula: C11H10FeO2 Formula Weight: 230.05 Melting Point: ca 210° dec. MDL No.: MFCD00001430

Formel: C₁₁H₁₀FeO₂
MW: 230,05 g/mol
Smältpunkt: ∼ 210 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00001430
CAS nummer: 1271-42-7
EINECS: 215-040-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR