1,1'-Ferrocenedicarboxylic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,1'-Dicarboxyferrocene
A16691.03 A16691.06 A16691.14
A16691.03EA 1975 SEK
A16691.03 A16691.06 A16691.14
1,1'-Ferrocenedicarboxylic acid ≥98%
1,1'-Ferrocenedicarboxylic acid
Purity: 98% CAS No.: 1293-87-4 Molecular Formula: C12H10FeO4 Formula Weight: 274.06 Odour: Odourless Melting Point: 300° MDL No.: MFCD00001423
Formel: C₁₂H₁₀FeO₄
MW: 274.06 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00001423
CAS nummer: 1293-87-4
EINECS: 215-068-9