Sodium hydroxymethanesulphinate, hydrate tech. 85% cont. up to 5% sodium sulfite, Technical Grade
Leverantör: Thermo Scientific
Varning
Synonyms: Sodium formaldehydesulphoxylate
A15731.36 A15731.0E
A15731.36EA 795 SEK
A15731.36 A15731.0E
Sodium hydroxymethanesulphinate, hydrate tech. 85% cont. up to 5% sodium sulfite, Technical Grade
Sodium hydroxymethanesulphinate
Purity: 85% CAS No.: 149-44-0 Molecular Formula: HOCH2SO2Na.xH2O Formula Weight: 118.09(anhy) Melting Point: 64-68° Beilstein Reference: 3630814 Merck Reference: 14,8620 MDL No.: MFCD00150598
Formel: CH₃NaO₃S·H₂O
MW: 118,09 g/mol
Smältpunkt: 64…68 °C
Storage Temperature: Omgivande
MDL Number: MFCD00150598
CAS nummer: 149-44-0
EINECS: 205-739-4
Merck Index: 14,8620