3-Methyl-1-phenyl-1H-pyrazole ≥98%
Leverantör: Thermo Scientific
Varning
A14841.03 A14841.09 A14841.14
A14841.03EA 615 SEK
A14841.03 A14841.09 A14841.14
3-Methyl-1-phenyl-1H-pyrazole ≥98%
3-Methyl-1-phenyl-1H-pyrazole
Purity: 98% CAS No.: 1128-54-7 Molecular Formula: C10H10N2 Formula Weight: 158.20 Flash Point: 110°(230°F) Odour: Odourless Beilstein Reference: 117870 MDL No.: MFCD00003123
Formel: C₁₀H₁₀N₂
MW: 158,2 g/mol
Smältpunkt: 35…38 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003123
CAS nummer: 1128-54-7
EINECS: 214-438-7