Dimethyl phenylphosphonite ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Phenyldimethoxyphosphine
A14006.03 A14006.06 A14006.14
A14006.03EA 645 SEK
A14006.03 A14006.06 A14006.14
Dimethyl phenylphosphonite ≥98%
Dimethyl phenylphosphonite
Purity: 98% CAS No.: 2946-61-4 Molecular Formula: C6H5P(OCH3)2 Formula Weight: 170.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5290 Beilstein Reference: 1073018 MDL No.: MFCD00008352
Formel: C₈H₁₁O₂P
MW: 170.15 g/mol
Densitet: 1.072
Flampunkt: >110 °C (230 °F)
MDL Number: MFCD00008352
CAS nummer: 2946-61-4
EINECS: 220-960-6