3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone
A11161.0E A11161.22 A11161.36
A11161.0EEA 5175 SEK
A11161.0E A11161.22 A11161.36
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98%
3-Methyl-1-phenyl-2-pyrazolin-5-one
Purity: 98+% CAS No.: 89-25-8 Molecular Formula: C10H10N2O Formula Weight: 174.20 Odour: Odourless Melting Point: 127-131° Boiling Point: 191°/17mm Beilstein Reference: 609575 Merck Reference: 14,6713 MDL No.: MFCD00003138
Formel: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Kokpunkt: 191 °C (17 mmHg)
Smältpunkt: 127…131 °C
Densitet: 1.120
Storage Temperature: Omgivande
MDL Number: MFCD00003138
CAS nummer: 89-25-8
EINECS: 201-891-0