Phenyl dichlorophosphate 97%, stavar

Leverantör: Thermo Scientific
Fara

Synonyms: Phenyl phosphorodichloridate

A10479.14 A10479.22 A10479.36
A10479.14EA 805 SEK
A10479.14 A10479.22 A10479.36
Phenyl dichlorophosphate 97%, stavar
Phenyl dichlorophosphate

Purity: 97% CAS No.: 770-12-7 Molecular Formula: C6H5Cl2O2P Formula Weight: 210.98 Flash Point: 112°(233°F) Refractive Index: 1.5230 Melting Point: -1° Boiling Point: 241-243° Beilstein Reference: 511863 UN No.: 3265 MDL No.: MFCD00002067

Formel: C₆H₅Cl₂O₂P
MW: 210,98 g/mol
Kokpunkt: 241…243 °C
Smältpunkt: –1 °C
Densitet: 1,415
Flampunkt: 112 °C (233 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002067
CAS nummer: 770-12-7
EINECS: 212-220-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR