Pentaethylene glycol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol
A10133.09 A10133.18 A10133.30
A10133.09EA 2875 SEK
A10133.09 A10133.18 A10133.30
Pentaethylene glycol ≥98%
Pentaethylene glycol
Purity: 98+% CAS No.: 4792-15-8 Molecular Formula: H(OCH2CH2)5OH Formula Weight: 238.28 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 338-340° Beilstein Reference: 1635593 MDL No.: MFCD00002878
Formel: C₁₀H₂₂O₆
MW: 238,28 g/mol
Kokpunkt: 338…340 °C
Densitet: 1,126
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002878
CAS nummer: 4792-15-8
EINECS: 225-341-4