(tert-Butyldimethylsilyl)acetylene 97%
Leverantör: Thermo Fisher Scientific
Fara
381300010 381300050
ACRO381300010EA 1250 SEK
ACRO381300010 ACRO381300050
(tert-Butyldimethylsilyl)acetylene 97%
(tert-Butyldimethylsilyl)acetylene
Formel: C₈H₁₆Si
MW: 140.3 g/mol
Flampunkt: 6 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191877
CAS nummer: 86318-61-8
UN: 1993
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %