2-Bromoethyl ethyl ether 90%
Leverantör: Thermo Fisher Scientific
Fara
367400250 367401000
ACRO367400250EA 1200 SEK
ACRO367400250 ACRO367401000
2-Bromoethyl ethyl ether 90%
2-Bromoethyl ethyl ether
Formel: C₄H₉BrO
Kokpunkt: 125…127 °C
Densitet: 1,357 g/cm³
Flampunkt: 21 °C
Storage Temperature: Frys
MDL Number: MFCD00000237
CAS nummer: 592-55-2
EINECS: 209-763-6
UN: 2340
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4435 to 1.4515 (20°C, 589 nm)
GC ≥88 %