Dichlorodiethylsilane 97%
Leverantör: Thermo Scientific
Fara
338100050 338100250
ACRO338100050EA 735 SEK
ACRO338100050 ACRO338100250
Dichlorodiethylsilane 97%
Dichlorodiethylsilane
Formel: C₄H₁₀Cl₂Si
MW: 157,11 g/mol
Smältpunkt: –96 °C
Flampunkt: 28 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000493
CAS nummer: 1719-53-5
EINECS: 217-005-0
UN: 1767
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4320 (20°C, 589 nm)
GC ≥96.0 %