Diethyl acetylenedicarboxylate 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Diethyl but-2-yne-1,4-dioate
296350250 296351000
ACRO296350250EA 1425 SEK
ACRO296350250 ACRO296351000
Diethyl acetylenedicarboxylate 97%
Diethyl acetylenedicarboxylate
Formel: C₈H₁₀O₄
Kokpunkt: 107…110 °C (11 mmHg)
Smältpunkt: 1…3 °C
Flampunkt: 94 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009186
CAS nummer: 762-21-0
EINECS: 212-095-8
UN: 1760
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
GC ≥96.0 %