Methacrolein 90% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Acrolein
Stabilization: med hydrokinon
292831000 292835000 292830100 292830500
ACRO292831000EA 11450 SEK
ACRO292831000 ACRO292835000 ACRO292830100 ACRO292830500
Methacrolein 90% stabilised
Methacrolein
Formel: C₄H₆O
MW: 70,09 g/mol
Kokpunkt: 69 °C (760 mmHg)
Smältpunkt: –81 °C
Densitet: 0,847 g/cm³
Flampunkt: –15 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006974
CAS nummer: 78-85-3
EINECS: 201-150-1
UN: 2396
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥88 %
Stabilizer 0.1 to 0.5 % Hydroquinone