Natrium-3-nitrobenzensulfonat 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt
209120010 209120025
ACRO209120010EA 1800 SEK
ACRO209120010 ACRO209120025
Natrium-3-nitrobenzensulfonat 99%
Natrium-3-nitrobenzensulfonat
Formel: C₆H₄NNaO₅S
MW: 225,16 g/mol
Smältpunkt: 350 °C
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007490
CAS nummer: 127-68-4
EINECS: 204-857-3
Order Now
Specifikation testresultat
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
pH 6 to 10 (1 % in water)