Tetraethylene glycol 99.5%
Leverantör: Thermo Fisher Scientific
Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol
149590010 149590025 149590100
ACRO149590010EA 1475 SEK
ACRO149590010 ACRO149590025 ACRO149590100
Tetraethylene glycol 99.5%
Tetraethylene glycol
Formel: C₈H₁₈O₅
MW: 194.23 g/mol
Kokpunkt: 324…330 °C (760 mmHg)
Smältpunkt: –4.1 °C
Densitet: 1.12 g/cm³
Flampunkt: 176 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002879
CAS nummer: 112-60-7
EINECS: 203-989-9
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
GC ≥99.4 %