Phenylphosphonic dichloride 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide
130802500 130800010
ACRO130802500EA 2400 SEK
ACRO130802500 ACRO130800010
Phenylphosphonic dichloride 97%
Phenylphosphonic dichloride
Formel: C₆H₅Cl₂OP
MW: 194,98 g/mol
Kokpunkt: 258 °C (760 mmHg)
Smältpunkt: 3 °C
Densitet: 1,39 g/cm³
Flampunkt: 204 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002070
CAS nummer: 824-72-6
EINECS: 212-534-3
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5600 (20°C, 589 nm)
Titration Argentometric ≥96.0 %